LabServ MicroSpin 21R (230V)
Product Image
LabServ MicroSpin 21R (230V)
Have Questions?
Catalog number LSCMS21R-230
Price (MYR)
12,300.00
Special offer
Ends: 31-Dec-2025
23,671.00
Save 11,371.00 (48%)
EA
Add to cart
Price (MYR)
12,300.00
Special offer
Ends: 31-Dec-2025
23,671.00
Save 11,371.00 (48%)