Product Image
MicroSpin 21 Microcentrifuge
This Microcentrifuge is applicable for high-speed usage in labs.
Have Questions?
Catalog NumberCapacity
LSCMS21-23036 x 1.5/2.2 mL
Catalog number LSCMS21-230
Price (MYR)
10,664.85
Each
Capacity:
36 x 1.5/2.2 mL
Price (MYR)
10,664.85
Each
  • Compact, low-noise centrifuge with precise temperature control, compatible with various rotors, ideal for high-speed use in research labs.
Specifications
Capacity36 x 1.5/2.2 mL
Max. Speed16,500 rpm
Max. RCF21,532 x g
RefrigeratedNo