CentriVap Mobile System
CentriVap Mobile System
Actual product may vary
Labconco
CentriVap Mobile System
CentriVap Mobile Systems combine the separate vacuum concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to laboratory. A 12-13 mm Rotor and Solvent Trap are also included. The interior can accommodate a vacuum pump.
Have Questions?
Catalog NumberElectrical Requirements
LBC7812035230 V 50 Hz 8 A
Catalog number LBC7812035
Price (MYR)
0.00
Each
Add to cart
Electrical Requirements:
230 V 50 Hz 8 A
Price (MYR)
0.00
Each
Add to cart
CentriVap Mobile Systems combine the separate vacuum concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to laboratory. A 12-13 mm Rotor and Solvent Trap are also included. The interior can accommodate a vacuum pump — the only additional component necessary to begin processing. 230V, 50Hz, UK plug
Specifications
CertificationsUL 3101-1/61010-1, CE (European Community) conformity
Electrical RatingInternational
Shipping Weight (English)210 lb.
Shipping Weight (Metric)95.0 kg
Voltage0 to 300 V
Chamber MaterialCorrosion Resistant Steel
Electrical Requirements230 V 50 Hz 8 A
Length (English)14.1 in.
Height (English)38.2 in.
Height (Metric)97.0 cm
Length (Metric)35.8 cm
Plug TypeBritish (UK)
Width (Metric)70.9 cm
Width (English)27.9 in.
Dimensions (L x W x H)14.1 x 27.9 x 38.2 in. (35.8 x 70.9 x 97.0 cm)
Unit SizeEach