▸
Tubing
▸
Rubber and Silicone Tubing
▸
Replacement Freeze Dry Tubing
Labconco
Replacement Freeze Dry Tubing
Neoprene construction. Labconco™ Replacement Freeze Dry Tubing allows moisture from condensation to be emptied from the freeze dryer.
Have Questions?
Catalog Number
Overall Length (English)
LBC7645900
3 ft.
Catalog number
LBC7645900
Price (MYR)
522.00
Each
Add to cart
Overall Length (English):
3 ft.
Price (MYR)
522.00
Each
Add to cart
For Dry Ice Benchtop Freeze Dry System connections or as drain hose on FreeZone 6, 12, and 18 Liter Freeze Dryers manufactured before August 2017
Replacement drain hose allows moisture from condensation to be emptied from the freeze dryer
8mm (5/16 in.) ID
Length: 0.9m (3 ft.)
Verify that the part matches your FreeZone Freeze Dryer's volume before purchasing
Specifications
Material
Rubber
Overall Length (English)
3 ft.
Flexibility
Semirigid