▸
Freeze Dryers
▸
Drying Equipment Accessories
▸
Fast-Freeze™ Flask Adapter
Actual product may vary
Labconco
Fast-Freeze™ Flask Adapter
Fast-Freeze™ Flask Adapter for connecting Fast-Freeze™ flasks to freeze-dryer valves.
Have Questions?
Change view
Catalog Number
Diameter (English)
Material
Style
LBC7547200
0.75 in.
Stainless Steel
Straight
LBC7545800
0.75 in.
Borosilicate Glass
45° bend
LBC7547600
0.75 in.
Stainless Steel
45° bend
3 Options
Catalog number
LBC7547200
Price (MYR)
308.00
Each
Add to cart
Diameter (English):
0.75 in.
Material:
Stainless Steel
Style:
Straight
Price (MYR)
308.00
Each
Add to cart
For connecting Fast-Freeze™ flasks to freeze-dryer valves
40 and 80 mL flasks take size 0.5 in. or 0.5 to 0.75 in.
Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications
Type
Flask Adapter
Capacity (Metric)
40, 80 mL
Material
Stainless Steel
Style
Straight
For Use With (Application)
For connecting fast freeze™ flasks to freeze-dryer valves
For Use With (Equipment)
Fast freeze™ flask
Diameter (English)
0.75 in.
Diameter (Metric)
1.9 cm
Height (English)
0.75 in.
Height (Metric)
1.905 cm
Length (English)
3.25 in.
Length (Metric)
8.26 cm
Weight (English)
0.5 lb.
Weight (Metric)
0.2 kg