Paramount™ Ductless Enclosure Filters
Paramount™ Ductless Enclosure Filters
Actual product may vary
Labconco
Paramount™ Ductless Enclosure Filters
Filters for Paramount Ductless Enclosures
Have Questions?
Catalog NumberFilter TypeFor Use With (Application)
LBC6938200CarbonOrganic Vapor #7.5
Catalog number LBC6938200
Price (MYR)
2,559.00
Each
Add to cart
Filter Type:
Carbon
For Use With (Application):
Organic Vapor #7.5
Price (MYR)
2,559.00
Each
Add to cart
For use with Paramount Ductless Enclosures
Specifications
Filter TypeCarbon
For Use With (Equipment)Paramount Ductless Enclosure
Height (English)1.5 in.
Weight (English)10 lb.
Weight (Metric)4.5 Kg
Length (English)10 to 20 in.
Depth (English)15 in.
Width (English)18 in.
Height (Metric)3.8 cm
Dimensions10 to 20 x 18 x 1.5 in. (25.4 to 50.8 x 45.7 x 3.8 cm)
Depth (Metric)38.1 cm
Length (Metric)25.4 to 50.8 cm
Width (Metric)45.7 cm
Product TypeFilter
Length (English) Exterior10 to 20 in.
For Use With (Application)Organic Vapor #7.5
Height Overall (English)1.5 in.
Height Overall (Metric)3.8 cm
Unit SizeEach