Specifications
DescriptionUpper Spindle Rack, FlaskScrubber and FlaskScrubber Vantage
MaterialStainless Steel
Product TypeRack
Rack TypeUpper Spindle Rack, FlaskScrubber and FlaskScrubber Vantage
For Use With (Equipment)Labconco FlaskScrubber or FlaskScrubber Vantage
Depth (English)21.6 in.
Depth (Metric)54.9 cm
Height (English)6.9 in.
Height (Metric)17.5 cm
Length (English)21.6 in.
Length (Metric)54.9 cm
Width (English)20.8 in.
Width (Metric)52.8 cm
Weight (English) Shipping23 lb.
Weight (Metric) Shipping10.4 kg
Quantity1/Ea.
Unit SizeEach