Glassware Washer Base Stand
Glassware Washer Base Stand
Actual product may vary
Labconco
Glassware Washer Base Stand
Accessories for use with Labconco Glassware Washers with CleanWorks OS.
Have Questions?
Catalog NumberDescription
LBC4653500FlaskScrubber and SteamScrubber Washer Base Stand
Catalog number LBC4653500
Price (MYR)
9,918.00
Each
Add to cart
Description:
FlaskScrubber and SteamScrubber Washer Base Stand
Price (MYR)
9,918.00
Each
Add to cart
  • Racks and accessories for use with Labconco FlaskScrubber, SteamScrubber, FlaskScrubber Vantage Glassware Washers
  • Base Stands elevate freestanding SteamScrubber and FlaskScrubber or FlaskScrubber Vantage washers 17 in. for loading convenience
  • Includes one storage compartment with hinged door
  • Type 304 stainless steel
  • Stationary stand, fully assembled with leveling feet
Specifications
DescriptionFlaskScrubber and SteamScrubber Washer Base Stand
MaterialStainless Steel
Product TypeBase Stand
Holds1 washer
For Use With (Equipment)Labconco FlaskScrubber or SteamScrubber
Depth (English)24.1 in.
Depth (Metric)61.2 cm
Height (English)17 in.
Height (Metric)43.2 cm
Length (English)24.1 in.
Length (Metric)61.2 cm
Width (English)25.4 in.
Width (Metric)64.5 cm
Weight (English) Shipping80 lb.
Weight (Metric) Shipping36.3 kg
Quantity1/Ea.
Unit SizeEach