Glassware Washer Dispenser
Actual product may vary
Labconco
Glassware Washer Dispenser
Accessories for use with Labconco Glassware Washers with CleanWorks OS.
Have Questions?
Catalog NumberDescription
LBC4587520Liquid Detergent Dispenser
Catalog number LBC4587520
Price (MYR)
7,897.00
Each
Description:
Liquid Detergent Dispenser
Price (MYR)
7,897.00
Each
  • Racks and accessories for use with Labconco FlaskScrubber, SteamScrubber, FlaskScrubber Vantage Glassware Washers.
  • Liquid Detergent Dispenser connects to any SteamScrubber or FlaskScrubber Washer to automate liquid detergent dispensing. Includes peristaltic pump, float switch, tubing, and one-gallon plastic container. Detergent not included.
  • Acid Rinse Dispenser connects to any SteamScrubber or FlaskScrubber Washer to automate acid neutralizing rinse aid dispensing. Includes peristaltic pump, float switch, tubing, and a one-liter plastic container. Rinse aid not included.
  • Detergent & Acid Rinse Dispenser connects to any SteamScrubber or FlaskScrubber Washer to automate liquid detergent & acid rinse aid dispensing on washers with 9-digit catalog number. Includes 1 gal detergent & 1 L rinse aid containers. Detergent & rinse aid not included.
Specifications
DescriptionLiquid Detergent Dispenser
MaterialPlastic
Product TypeWasher Dispenser
ColorWhite
For Use With (Equipment)Labconco FlaskScrubber or SteamScrubber
Depth (English)6 in.
Depth (Metric)15.2 cm
Height (English)11.8 in.
Height (Metric)30.0 cm
Length (English)6 in.
Length (Metric)15.2 cm
Width (English)6 in.
Width (Metric)15.2 cm
Weight (English) Shipping3 lb.
Weight (Metric) Shipping1.4 kg
Quantity1/Ea.
Unit SizeEach