Kimtech™ Pure G3 Cleanroom NXT™ Nitrile Gloves
Actual product may vary
Ansell
Kimtech™ Pure G3 Cleanroom NXT™ Nitrile Gloves
Non-sterile, nitrile gloves do not contain natural rubber latex proteins. Recommended for ISO Class 4 or higher cleanroom environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC62992Medium
KLC62990X-Small
KLC62991Small
KLC62993Large
Catalog number KLC62992
Price (MYR)
1,821.00
Case
Size:
Medium
Price (MYR)
1,821.00
Case
Non-sterile nitrile gloves do not contain natural rubber latex proteins. Recommended for ISO Class 4 or higher cleanroom environments.
  • Textured fingertips
  • Beaded cuff for easy donning
  • Contain no natural rubber latex
  • Ambidextrous
  • Thickness: 6.2 mil at fingertips
  • Length: 12 in. (30.5 cm)
  • White
  • Meet AQL level of 1.5 for pinholes
  • Packaging: 10 bags of 100
Specifications
Certifications/ComplianceASTM D6978-05: Resistance to permeation by chemo drugs
Cleanroom ClassClass 10 (ISO 4)
AQL1.5
ColorWhite
Exterior FinishPartially Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
PowderedPowder Free
SizeMedium
SterilityNon-sterile
Thickness (English)6 mil
Thickness (Metric)0.15 mm
Model62992
Quantity10 Bags of 100
SeriesKimtech™ G3
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase