KleenGuard™ A40 Reflex™ Liquid and Particle Protection Coveralls
Actual product may vary
Ansell
KleenGuard™ A40 Reflex™ Liquid and Particle Protection Coveralls
Coveralls feature a respirator fit hood, zipper front, elastic wrists and ankles. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
Have Questions?
Catalog NumberSize
KLC47996Large
Catalog number KLC47996
Price (MYR)
-
Size:
Large
  • Feature respirator fit hood, zipper front, elastic wrists and ankles
  • Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates
  • Pass NFPA 99 for antistatic materials
  • Pass ASTM F1670 testing for penetration of blood and bodily fluids
  • Reflex sizing for a more comfortable fit and less rip outs
  • Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing
Specifications
Product TypeGeneral Purpose Coveralls
SizeLarge
GenderUnisex
MaterialMicroporous Film Laminate
ColorWhite
DisposableLimited-use
Ankle StyleElastic
Cuff StyleElastic
Neck StyleHooded
Hood TypeElastic, Respirator Fit
Closure TypeFront Zipper
Seam TypeSerged
SterilityNon-sterile
ResistanceLiquid, Particle, Anti-static, Chemicals, Bloodborne Pathogens
Certifications/ComplianceNFPA 99, ASTM F1670
ModelKleenGuard™ A40 Reflex™
No. of PocketsNone
For Use With (Application)Aviation, Pharmaceutical Manufacturing, General Manufacturing, Liquid Handling
Unit SizeCase