A8 Clean Overalls
Actual product may vary
Ansell
A8 Clean Overalls
Maintain ISO 7 Class C or higher cleanroom environments with breathable, ultra light overalls. Kimtech™ A8 Clean Overalls are made of a high performance, chemical resistant fabric and come in compressed packaging to save space in the gowning room.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC47693Large
KLC47691Small
KLC47692Medium
KLC47694X-Large
KLC476952X-Large
Catalog number KLC47693
Price (MYR)
605.00
Case
Size:
Large
Price (MYR)
605.00
Case
  • High performance chemical resistant fabric
  • Innovative, ultra light fabric: 45 gsm
  • Breathable
  • Highly comfortable
  • Compressed packaging to save space in the gowning room
Specifications
Product TypeCleanroom Coveralls
SizeLarge
MaterialSMS
ColorWhite
DisposableSingle-use
Waist StyleElastic
Ankle StyleElastic
Cuff StyleThumb Loop
Neck StyleCollared
Hood TypeUnhooded
Closure TypeZipper
Seam TypeStitched
SterilityNon-sterile
ResistanceNone
Collar TypeStanding
Certifications/CompliancePPE Cat III Type 5/6 (Protection against particles/limited splash protection), EN1149-1:1995 Antistatic garment
For Use With (Application)Controlled Environments
Unit SizeCase