Kimtech Prep™ Kimtex™ Wipers
Actual product may vary
Ansell
Kimtech Prep™ Kimtex™ Wipers
Heavy-duty wiper for oil and solvent cleanups
Have Questions?
Catalog NumberDimensions (L x W)
KLC3356013 x 12.5 in. (33 x 31.75 cm)
Catalog number KLC33560
Price (MYR)
-
Dimensions (L x W):
13 x 12.5 in. (33 x 31.75 cm)
  • Reusable wiper is excellent for cleaning and prep on surfaces and parts and for glass cleaning
  • Constructed of low-lint polypropylene for long-lasting performance
  • Absorb liquids quickly — ideal for big cleanups
  • Acid-, base-, and solvent-resistant
  • Blue
Specifications
Product TypeHeavy-duty Wiper
Length (English)13 in.
Length (Metric)33 cm
Width (English)12 in.
Width (Metric)31.75 cm
MaterialPolypropylene
ColorBlue
DisposableReusable
Dimensions (L x W)13 x 12.5 in. (33 x 31.75 cm)
For Use With (Application)General-Purpose
Unit SizeCase