G3 Sterile Sterling™ Nitrile Cleanroom Gloves
Actual product may vary
G3 Sterile Sterling™ Nitrile Cleanroom Gloves
G3 Sterile Sterling™ Nitrile Cleanroom Gloves
G3 Sterile Sterling™ Nitrile Cleanroom Gloves
G3 Sterile Sterling™ Nitrile Cleanroom Gloves
G3 Sterile Sterling™ Nitrile Cleanroom Gloves
Ansell
G3 Sterile Sterling™ Nitrile Cleanroom Gloves
With high levels of contamination protection, Kimtech™ G3 Sterling™ sterile nitrile gloves are used in EU GMP ISO Class 5 Grade A or higher sterile cleanrooms. The hand-specific gloves are suitable for double donning, latex-free, and non-pyrogenic.
Have Questions?
Catalog NumberSize
KLC118226.5
Catalog number KLC11822
Price (MYR)
2,120.00
Case
Size:
6.5
Price (MYR)
2,120.00
Case
G3 Sterile Sterling™ nitrile gloves provide protection and cleanliness for Class 10/ISO 4 cleanroom environments.
Assured Compliance:
  • PPE Cat III according to Regulation (EU) 2016/425
  • EN ISO 374-1:2016 Type C (K) Chemical Splash protection
  • EN 374-4:2003 Resistance to degradation by chemicals
  • EN ISO 374-5:2016 Micro Organism and VIRUS Protection
  • ASTMD6978-05 cytotoxic drugs splash protection
  • Sterility Assurance Level (SAL) 10-6
  • Certificate of Analysis available online
  • Certificate of Sterility available online
  • Efficient, environmentally-friendly construction and cleanroom-ready packaging that minimizes waste without compromising safety.
  • The fully sterilized nitrile material is stronger and leaner than latex, and features certified protection against a wide range of contaminants, including viruses, micro-organisms and chemical splash.
  • Gloves are hand-specific, disposable and gray in color with a high level of cleanliness.
  • Textured fingertips enhance grip and tactile sensitivity for safer and more efficient processes.
  • Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
  • Contain no natural rubber latex, silicone or powder, reducing the risks of skin irritation for the wearer.
Specifications
Certifications/ComplianceASTM D6319, ASTM D3767
Certifications/Compliance (Europe)EN ISO 374-1:2016; EN 374-4:2003; EN ISO 374-5:2016; ASTMD6978-05
Cleanroom ClassClass 1 (ISO 3)
Accelerator FreeYes
AQL1.5
ColorGray
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)4 mil
Finger Thickness (Metric)0.10 mm
HandSpecific
Latex FreeYes
MaterialNitrile
Palm Thickness (English)3.1 mil
Palm Thickness (Metric)0.08 mm
PowderedPowder Free
Product TypeCleanroom Gloves
Size6.5
SterilitySterile
Thickness (English)4 mil
Thickness (Metric)0.10 mm
Manufacturing/QMS Audit StandardsISO 9001
Model11822
SeriesKimtech™ G3 Sterile
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase