Hygienic Wall Bracket, Single Hook Module
Hygienic Wall Bracket, Single Hook Module
Actual product may vary
Ansell
Hygienic Wall Bracket, Single Hook Module
This single hook module is designed for storing cleaning tools that have a hanging hole. The hook is slid onto the supplied single-base/spacer from either side.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
KLC10124Red
TYL10122Green
Catalog number KLC10124
Price (MYR)
531.00
Case
Add to cart
Color:
Red
Price (MYR)
531.00
Case
Add to cart
  • Single hook module can hold products weighing up to 3 kg
  • Easy to dissemble foeaning or replacement
  • FDA compliant raw material
Specifications
Product TypeWall Bracket
ColorRed
MaterialPolypropylene
Height (English)1.9 in.
Length (English)1.6 in.
Weight (English)0.06 in.
Width (English)3.1 in.
Unit SizeCase