Sure-Grip™ EX Corrosives/Acid Steel Safety Cabinet
Actual product may vary
Justrite
Sure-Grip™ EX Corrosives/Acid Steel Safety Cabinet
Justrite Sure-Grip™ EX Corrosives/Acid Steel Safety Cabinet offers safe and compliant storage options for corrosives.
Have Questions?
Catalog NumberDimensions (D x W x H) InteriorLoad Bearing Capacity
JUS89450214.56 x 39.5 x 60.5 in.45 gal. (170 L)
Catalog number JUS894502
Price (MYR)
-
Dimensions (D x W x H) Interior:
14.56 x 39.5 x 60.5 in.
Load Bearing Capacity:
45 gal. (170 L)
  • Features double-wall construction, dual vents, grounding wire connections
  • Adjustable shelves, leakproof sills, three point self-latching doors and leveling feet
  • Acid cabinets contains polyethylene trays attached to galvanized steel shelves to resist aggressive chemicals
  • Polyethylene liner at bottom sump
  • Epoxy baked-on powder coat finish
  • Paint resists acids, bases and solvents
Specifications
Certifications/ComplianceFM, NFPA, OSHA
Depth (English) Interior14.56 in.
Depth (Metric) Exterior45.72 cm
Depth (Metric) Interior37 cm
DescriptionLiner: Polyethylene
Dimensions (D x W x H) Interior14.56 x 39.5 x 60.5 in.
Door StyleManual-close
For Use With (Application)Corrosives
Height (English) Interior60.5 in.
Height (Metric) Exterior165.1 cm
Height (Metric) Interior153.7 cm
IncludesDouble wall, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self latching doors and leveling feet, Polyethylene trays, separate Polyethylene liner
Load Bearing Capacity45 gal. (170 L)
No. of Doors2
No. of Shelves2
Surface CoatingEpoxy
Weight (English)301 lb.
Weight (Metric)136.5 kg
Width (English) Interior39.5 in.
Width (Metric) Exterior109 cm
Width (Metric) Interior100.3 cm
ColorBlue
Depth (English) Exterior18 in.
Height (English) Exterior65 in.
MaterialSteel (CR)
Product TypeCorrosives and Acids Safety Cabinet
Width (English) Exterior43 in.
Unit SizeEach