FlexiPump Dispensing Assembly
Actual product may vary
Interscience
FlexiPump Dispensing Assembly
Dispensing assembly for use with FlexiPump.
Have Questions?
Catalog NumberDiameter (Metric) Inner
ISC5611023.2 mm
Catalog number ISC561102
Price (MYR)
2,415.00
Each
Diameter (Metric) Inner:
3.2 mm
Price (MYR)
2,415.00
Each
Includes:
  • Set of silicone tubings
  • GL45 screw cap
  • Tube weight
  • Nozzle
  • Cap
  • Filter
Choose between tubing inner diameter of 1.6 mm, 3.2 mm, 4.8 mm, 6.4 mm, or 8 mm
Specifications
AutoclavableAutoclavable
Diameter (English) Inner0.12 in.
Diameter (Metric) Inner3.2 mm
MaterialSilicone
Thickness (English)0.12 in.
For Use WithFlexiPump
SterilityNon-sterile
ColorClear
Product TypeTubing Assembly
Unit SizeEach