Interchangeable Block for QB Series Dry Block Heaters
Actual product may vary
Grant
Interchangeable Block for QB Series Dry Block Heaters
For use with 0.2mL tubes, strips and 96-well microtitre plates used in molecular biology and biotechnology applications. Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters can withstand temperatures up to 100°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTube SizeNo. of PlacesDepth (Metric)
GRAQB-1212mm2450 mm hole depth
GRAQB-17H17mm1075 mm hole depth
GRAQB-1313mm1250 mm hole depth
FKS1143091816mm1250 mm hole depth
GRAQB-1010mm2450 mm hole depth
Catalog number GRAQB-12
Price (MYR)
1,051.00
Each
Tube Size:
12mm
No. of Places:
24
Depth (Metric):
50 mm hole depth
Price (MYR)
1,051.00
Each
  • For use with QBD1, QBD2, QBD4, QBH2, QBA1 and QBA2 dry block heating system
  • Accommodates a variety of test tube sizes
  • Available in plain block format without holes
Specifications
For Use With (Equipment)Test Tubes (12 mm)
MaterialAluminum
No. of Places24
TypeBlock
Tube Size12mm
Dimensions (L x W x H)140 x 50 x 63 mm
Depth (Metric)50 mm hole depth
Unit SizeEach