▸
Shakers
▸
Rockers and Rotators
▸
Universal Platform with Adjustable Bars
Actual product may vary
Grant
Universal Platform with Adjustable Bars
Adjust bars to fit a variety of vessels at once. Grant Instruments™ Universal Platform and Adjustable Bars does not require any tools to adjust bars.
Have Questions?
Catalog Number
For Use With (Equipment)
GRAPUP-12
ES-20
Catalog number
GRAPUP-12
Price (MYR)
1,264.00
Each
Add to cart
For Use With (Equipment):
ES-20
Price (MYR)
1,264.00
Each
Add to cart
For use with Grant Orbital Shakers and Shaker Incubators
Dimensions: 270 x 195mm
Includes Extra Holding Bars which can also be ordered separately (HB 200)
Specifications
Product Type
Universal Platform
Dimensions (L x W)
285 x 220 mm
For Use With (Application)
For Different Types Of Container
For Use With (Equipment)
ES-20