Buffer Solution pH 10.01
Buffer Solution pH 10.01
Actual product may vary
Buffer Solution pH 10.01
Buffers and calibration solutions packed in durable plastic bottles – economical and convenient for laboratory use. ECBU10BTC
Have Questions?
Catalog number FSP01X211205
Price (MYR)
73.00
Special offer
94.00
Save 21.00 (22%)
EA
Add to cart
Price (MYR)
73.00
Special offer
94.00
Save 21.00 (22%)
EA
Add to cart
Buffers and calibration solutions packed in durable plastic bottles – economical and convenient for laboratory use. ECBU10BTC
  • Buffers and calibration solutions
  • Packed in 480 mL durable plastic bottles
  • Economical and convenient for laboratory use
Specifications
CAS497-19-8
Boiling PointApproximately 100°C
ColorBlue
CAS3844-45-9
CAS144-55-8
For Use With (Application)Measuring pH
CAS99-76-3
For Use With (Equipment)pH Meter
pH10
Physical FormLiquid
Solution TypeAqueous
CAS7732-18-5
FormulationSodium carbonate, Deionized water, Methyl Paraben, Erioglaucine
PackagingPlastic Bottle
Unit SizeEA