L-Theanine, 98%
L-Theanine, 98%
L-Theanine, 98%
Thermo Scientific Chemicals
L-Theanine, 98%
CAS: 3081-61-6 | C7H14N2O3 | 174.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA459340050
Price (MYR)
939.00
Each
Quantity:
5 g
Request bulk or custom format
Price (MYR)
939.00
Each
Specifications
Chemical Name or MaterialL-Theanine
CAS3081-61-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee