Bromelain, sourced from stem, >=2500 GDU/g (>=5.0 FIP U/mg)
Thermo Scientific Chemicals
Bromelain, sourced from stem, >=2500 GDU/g (>=5.0 FIP U/mg)
CAS: 37189-34-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA448210250
Price (MYR)
537.00
Each
Quantity:
25 g
Request bulk or custom format
Price (MYR)
537.00
Each
Specifications
Chemical Name or MaterialBromelain
Name Note2500 GDU/g
CAS37189-34-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.