Basic Fuchsin, 88%, pure
Basic Fuchsin, 88%, pure
Basic Fuchsin, 88%, pure
Thermo Scientific Chemicals
Basic Fuchsin, 88%, pure
CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA419141000100 gGlass bottle
Catalog number FSA419141000
Price (MYR)
1,758.00
Bottle
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,758.00
Bottle
Specifications
Absorbance1800 (at lambda min.)
Chemical Name or MaterialBasic Fuchsin
Melting Point250.0°C
Name Note88%
CAS632-99-5