Iron(II) chloride, 97%, anhydrous
Iron(II) chloride, 97%, anhydrous
Iron(II) chloride, 97%, anhydrous
Thermo Scientific Chemicals
Iron(II) chloride, 97%, anhydrous
CAS: 7758-94-3 | Cl2Fe | 126.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
Glass bottle
Catalog number FSA389352500
Price (MYR)
3,781.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,781.00
Each
Specifications
Boiling Point1026.0°C
Chemical Name or MaterialIron(II) chloride
Melting Point677.0°C
CAS7758-94-3
ColorBrown