Calcium chloride hexahydrate, 98+%, for analysis
Calcium chloride hexahydrate, 98+%, for analysis
Calcium chloride hexahydrate, 98+%, for analysis
Thermo Scientific Chemicals
Calcium chloride hexahydrate, 98+%, for analysis
CAS: 7774-34-7 | CaCl2·6H2O | 219.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA389262500
Price (MYR)
234.00
Bottle
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
234.00
Bottle
Specifications
Chemical Name or MaterialCalcium chloride hexahydrate
Melting Point30.0°C
Name NoteFor analysis
CAS7774-34-7