▸
Organic compounds
▸
Organooxygen compounds
▸
Carbonyl compounds
▸
Complex Ketones
▸
4'-Fluoro-3'-nitroacetophenone, 98%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
4'-Fluoro-3'-nitroacetophenone, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number
FSA366010050
Price (MYR)
406.00
Gram
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
406.00
Gram
Add to cart
Specifications
Chemical Name or Material
4'-Fluoro-3'-nitroacetophenone
CAS
400-93-1
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0