Silver, 99.9%, (trace metal basis), wire, 0.5mm diam., length 10m, 1ea = 20gr
Silver, 99.9%, (trace metal basis), wire, 0.5mm diam., length 10m, 1ea = 20gr
Silver, 99.9%, (trace metal basis), wire, 0.5mm diam., length 10m, 1ea = 20gr
Fisherbrand
Silver, 99.9%, (trace metal basis), wire, 0.5mm diam., length 10m, 1ea = 20gr
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
FSA3177100101 Ea.
Catalog number FSA317710010
Price (MYR)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver
ColorSilver
Name Note99.9% (1ea = 20gr)
CAS7440-22-4
Melting Point961.0°C
View more
for ultra trace analysis
RUO – Research Use Only