Titanium(IV) oxide, 98.0-100.5% TiO2
Titanium(IV) oxide, 98.0-100.5% TiO2
Titanium(IV) oxide, 98.0-100.5% TiO2
Titanium(IV) oxide, 98.0-100.5% TiO2
Thermo Scientific Chemicals
Titanium(IV) oxide, 98.0-100.5% TiO2
CAS: 13463-67-7 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
Packaging:
View all
Plastic drum
Catalog number FSA277370100
Price (MYR)
2,756.00
Each
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
2,756.00
Each
Specifications
Boiling Point2900.0°C
Chemical Name or MaterialTitanium(IV) oxide
Melting Point1855.0°C
CAS13463-67-7
ColorWhite
View more
Suitable for battery materials development.
RUO – Research Use Only