Dowex™ 1X8, 50-100 mesh, ion-exchange resin
Dowex™ 1X8, 50-100 mesh, ion-exchange resin
Dowex™ 1X8, 50-100 mesh, ion-exchange resin
Fisherbrand
Dowex™ 1X8, 50-100 mesh, ion-exchange resin
Dowex 1X8, C29H34ClN, CAS Number-12627-85-9, dowex 1x8 50-100 cl, dowex 1x2 50-100 cl, 500g, White, 01,511; 04,267; 06,302; 07,182; 08,263, BBQMUEOYPPPODD-UHFFFAOYSA-M, 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride, MFCD00132718, 432.048, Plastic bottle | CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA202991000100 gPlastic bottle
Catalog number FSA202991000
Price (MYR)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 1X8
Physical FormPowder and/or Chunks
CAS12627-85-9
Particle Size15 % max. (on 35 mesh, cumulative) 15 % max. (through 80 mesh)
PackagingPlastic bottle
View more
Ion exchange resin
RUO – Research Use Only