Amberlite™ IRA-900, Cl-form, ion-exchange resin
Amberlite™ IRA-900, Cl-form, ion-exchange resin
Amberlite™ IRA-900, Cl-form, ion-exchange resin
Thermo Scientific Chemicals
Amberlite™ IRA-900, Cl-form, ion-exchange resin
CAS: 9050-97-9 | C4H12ClN | 109.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA202315000
Price (MYR)
483.00
Bottle
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
483.00
Bottle
Specifications
Chemical Name or MaterialAmberlite™ IRA-900
Physical FormBeads
CAS9050-97-9
Particle SizeParticle size analysis : uniformity coefficient: <1.8, harmonic mean size: 0.650 to 0.820mm, <0.300mm: <0.5%,> 1.180mm: <3.0%
PackagingPlastic bottle
View more
Ion exchange resin
RUO – Research Use Only