Tungsten(VI) chloride, 99%, 20 mesh
Tungsten(VI) chloride, 99%, 20 mesh
Tungsten(VI) chloride, 99%, 20 mesh
Thermo Scientific Chemicals
Tungsten(VI) chloride, 99%, 20 mesh
CAS: 13283-01-7 | Cl6W | 396.57 g/mol
Have Questions?
Catalog NumberQuantity
FSA20030050050 g
Catalog number FSA200300500
Price (MYR)
1,084.00
Gram
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,084.00
Gram
Specifications
Boiling Point347°C
Chemical Name or MaterialTungsten(VI) chloride
Melting Point275°C
Name Note20 Mesh
CAS13283-01-7