Lithium perchlorate trihydrate, 99%, extra pure
Lithium perchlorate trihydrate, 99%, extra pure
Lithium perchlorate trihydrate, 99%, extra pure
Thermo Scientific Chemicals
Lithium perchlorate trihydrate, 99%, extra pure
CAS: 13453-78-6 | ClLiO4·3H2O | 160.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA194722500
Price (MYR)
1,165.00
Bottle
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,165.00
Bottle
Specifications
Chemical Name or MaterialLithium perchlorate trihydrate
Melting Point95°C
Name Notep.a.
CAS13453-78-6
ColorWhite
View more
Oxidation reagent
RUO – Research Use Only