Lithium chloride, 99%, extra pure
Lithium chloride, 99%, extra pure
Lithium chloride, 99%, extra pure
Thermo Scientific Chemicals
Lithium chloride, 99%, extra pure
CAS: 7447-41-8 | ClLi | 42.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number FSA193370010
Price (MYR)
2,333.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,333.00
Each
Specifications
Boiling Point1382.0°C
Chemical Name or MaterialLithium chloride
Melting Point605.0°C
CAS7447-41-8
Health Hazard 1GHS Signal Word: Warning