Magnesium, 99.9+%, (trace metal basis), turnings
Magnesium, 99.9+%, (trace metal basis), turnings
Magnesium, 99.9+%, (trace metal basis), turnings
Thermo Scientific Chemicals
Magnesium, 99.9+%, (trace metal basis), turnings
CAS: 7439-95-4 | Mg | 24.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number FSA191085000
Price (MYR)
431.00
Bottle
Quantity:
500 g
Request bulk or custom format
Price (MYR)
431.00
Bottle
Specifications
Chemical Name or MaterialMagnesium
Name Note99.9+%
CAS7439-95-4
Melting Point651.0°C
Boiling Point1107.0°C
View more
For ultra trace analysis
RUO – Research Use Only