Methacrylic acid, 99.5%, extra pure, stabilized
Methacrylic acid, 99.5%, extra pure, stabilized
Methacrylic acid, 99.5%, extra pure, stabilized
Thermo Scientific Chemicals
Methacrylic acid, 99.5%, extra pure, stabilized
CAS: 79-41-4 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
500 mL
1 L
Packaging:
View all
Glass bottle
Catalog number FSA168312500
Price (MYR)
265.00
Bottle
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
265.00
Bottle
Specifications
Chemical Name or MaterialMethacrylic acid
Name NoteStabilized
CAS79-41-4
Health Hazard 1SIGNAL WORD: DANGER!
Health Hazard 2GHS P Statement
May cause respiratory irritation.
Toxic in contact with skin.