L-Methionine, 98+%
L-Methionine, 98+%
L-Methionine, 98+%
Thermo Scientific Chemicals
L-Methionine, 98+%
CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic Bottle
Catalog number FSA166160250
Price (MYR)
322.00
Bottle
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
322.00
Bottle
Specifications
Chemical Name or MaterialL-Methionine
Melting Point276.0°C to 279.0°C
Name Note98+%
Ammonium300ppm max.
CAS63-68-3