Activated charcoal, NORIT™ A SUPRA
Activated charcoal, NORIT™ A SUPRA
Activated charcoal, NORIT™ A SUPRA
Thermo Scientific Chemicals
Activated charcoal, NORIT™ A SUPRA
Activated charcoal,NORIT™ A SUPRA, C, CAS Number-7440-44-0, acticarbone, activated charcoal, anthrasorb, carbon-12, carbono, carbosieve, graphene, graphite, mineral, norit, 1kg, Passes Test, CHEBI:27594, Black, 231-153-3, OKTJSMMVPCPJKN-UHFFFAOYSA-N, 15% max., 3652.0 deg.C, 15, 1807 | CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA134342500
Price (MYR)
730.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
730.00
Each
Specifications
Chemical Name or MaterialActivated charcoal,NORIT* A SUPRA
AcidityPasses Test
CAS7440-44-0
Melting Point3652.0°C
ColorBlack
View more
Filter aid
RUO – Research Use Only