Sulfur trioxide pyridine complex, technical, 48-50% active SO3
Sulfur trioxide pyridine complex, technical, 48-50% active SO3
Sulfur trioxide pyridine complex, technical, 48-50% active SO3
Thermo Scientific Chemicals
Sulfur trioxide pyridine complex, technical, 48-50% active SO3
CAS: 26412-87-3 | C5H5NO3S | 159.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA132871000
Price (MYR)
620.00
Bottle
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
620.00
Bottle
Specifications
Chemical Name or MaterialSulfur trioxide pyridine complex
Name Note48.8 to 50.3% active SO3
CAS26412-87-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
View more
Lewis acid
RUO – Research Use Only