▸
Cell Culture Utensils
▸
Inoculating Loops and Needles
▸
Twisted Nichrome™ Inoculating Loops with Handles
Actual product may vary
Twisted Nichrome™ Inoculating Loops with Handles
Available in three loop sizes
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inside
Gauge
FKS16406849
3 mm
24 G
FIS14-956-104
5 mm
24 G
2 Options
Catalog number
FKS16406849
Price (MYR)
-
Diameter (Metric) Inside:
3 mm
Gauge:
24 G
Available in three loop sizes
Aluminum handle is 7.25 in. long (18.4cm)
Supplied with nichrome wire loop
Length of wire is 100mm
Specifications
Diameter (English) Inside
0.118 in.
Diameter (Metric) Inside
3 mm
Material
Nichrome
Product Type
Inoculating Loop
Gauge
24 G
Length (English) Handle
7.25 in.
Length (Metric) Handle
18.4 cm
Handle Type
Aluminum handle