PTFE Magnetic Stir Bar, Cross-shaped
Actual product may vary
Fisherbrand
PTFE Magnetic Stir Bar, Cross-shaped
Use with containers, vessel, stirrers, and stirring hot plates. The PTFE-coated magnet stirrers are stable for many general purpose applications. The bars are cross-shaped for improved stirring action.
Have Questions?
Catalog NumberLength (Metric)
FKS1628579150 mm
Catalog number FKS16285791
Price (MYR)
345.00
Pack
Length (Metric):
50 mm
Price (MYR)
345.00
Pack
  • Stable, general-purpose magnetic stirrer
  • Provides a deep, quiet vortex even at relatively high speeds
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
Dimensions (L x W x H)50 x 50 x 15mm
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Height (Metric)15mm
Length (English)2 in.
Width (English)1 in.
Width (Metric)50mm
Length (Metric)50 mm
Quantity2/Pk.
ShapeCross
Unit SizePack