▸
Personal Protective Equipment
▸
Safety Clothing
▸
Aprons
▸
Shermond Plastic Aprons
Actual product may vary
Fisherbrand
Shermond Plastic Aprons
Shermond Plastic Aprons constructed of disposable polythene that feature a halter neck and waist ties.
Have Questions?
Catalog Number
Dimensions (L x W)
FKS12970245
117 x 69 cm
Catalog number
FKS12970245
Price (MYR)
-
Dimensions (L x W):
117 x 69 cm
Each roll is individually wrapped for product protection and ease of insertion into the wall mountable dispenser
PPE Class 1 offering protection from low level hazards where risk of injury is minimal
Store in a cool dry place out of direct sunlight
Specifications
Type
Apron
Material
Polythene
Color
White
Disposable
Single-use
Dimensions (L x W)
117 x 69 cm