▸
Clamps and Supports
▸
Clamps
▸
3-Prong Utility Clamp
Actual product may vary
Fisherbrand
3-Prong Utility Clamp
With a screwed shaft and nickel-plated bosshead which can be screwed up and down shaft 35mm and locked in any position with a wing nut.
Have Questions?
Catalog Number
Material
FKS12926826
Aluminum Nickel, Plastic
Catalog number
FKS12926826
Price (MYR)
598.00
Each
Add to cart
Material:
Aluminum Nickel, Plastic
Price (MYR)
598.00
Each
Add to cart
With jaws covered in chemically resistant plastic for a safe and secure hold
Bosshead accepts rods up to 13mm
Rods are secured with a stainless steel thumb screw
Specifications
Quantity
1 Ea.
Type
Utility Clamp
Material
Aluminum Nickel, Plastic
Size (Metric) Grip
90 mm Max.