Soda Lime Glass Petri Dishes
Actual product may vary
Fisherbrand
Soda Lime Glass Petri Dishes
High-quality petri dish, with lid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)
FKS12921408100 mm15 mm
FKS1290140860 mm12 mm
FKS12931408120 mm20 mm
FKS12941408150 mm25 mm
FKS1291140880 mm15 mm
Catalog number FKS12921408
Price (MYR)
932.00
Pack
Diameter (Metric):
100 mm
Height (Metric):
15 mm
Price (MYR)
932.00
Pack
  • Constructed of soda lime glass.
  • Includes lid
Specifications
Closure IncludedClosure Included
Closure MaterialSoda Lime Glass
MaterialSoda Lime Glass
ShapeRound
TypePetri Dish
Height (Metric)15 mm
Diameter (Metric)100 mm
Unit SizePack