Fisherbrand
Sharpsguard™ yellow 2.5 Multi-Purpose 2.21L Sharps Container
An ideal general-purpose sharps container. Sharpsguard™ yellow 2.5 Multi-Purpose 2.21L Sharps Container is for disposal of sharps that have been contaminated with medicinal products, but not cytotoxic or cytostatic.
Have Questions?
Catalog NumberColorDimensions (L x W x H)
FKS12913495Yellow176L x 45W x 163 mmH
Catalog number FKS12913495
Price (MYR)
-
Color:
Yellow
Dimensions (L x W x H):
176L x 45W x 163 mmH
  • Provides maximum capacity in the minimum working space
  • Container for disposal by incineration
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection against penetrating sharps and leaking fluids
  • When container is full to the fill line it must be locked and disposed of safely
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications
TypeStorage Container
MaterialPolypropylene
ColorYellow
Capacity (Metric)2.5 (Overall) L
Dimensions (L x W x H)176L x 45W x 163 mmH
Length (Metric)176 mm
Width (Metric)45 mm
Height (Metric)163 mm
Unit SizeEach