Vacuum Pump with UK, US, and Schuko Cables
Actual product may vary
Fisherbrand
Vacuum Pump with UK, US, and Schuko Cables
Quiet in operation with very low vibration. the Fisherbrand™ Vacuum Pump is designed for general duty laboratory applications requiring only rough vacuum or moderate pressure.
Have Questions?
Catalog NumberPump Type
FKS12911151Vacuum Pump
Catalog number FKS12911151
Price (MYR)
7,972.00
Each
Pump Type:
Vacuum Pump
Price (MYR)
7,972.00
Each
  • Compact filtration pump with small footprint
  • Economical to purchase and operate
  • Quiet running oil free pump
  • Lightweight and highly portable
  • DN 6/8 hose nozzle For vacuum filtration.
Specifications
Max. Vacuum100 mbar
Electrical Requirements90/260 V 50/60 Hz with Schuko UK and US power leads
Dimensions (L x W x H)14.4 x 12.7 x 19.6 cm (5.6 x 5 x 7.7 in.)
Weight (English)2 lb.
Weight (Metric)2.3 kg
For Use WithSchuko, UK And US Vacuum Cables
Pump TypeVacuum Pump
Air Displacement15 L/min.
ApplicationsAspiration
Maximum Pressure2.5 bar
Unit SizeEach