Transfer liquids safely and accurately with Thermo Scientific™ Sterilin™ Plastic Transfer Pipettes. Ideal for general laboratory use, sample storage and transport.
Have Questions?
Change view
Catalog Number
Volume (Metric)
Quantity
FKS12909487
3 mL
500/Pk., 3000/Cs.
FKS11369203
3 mL
10/Pk., 500/Cs.
2 Options
Catalog number FKS12909487
Price (MYR)
640.00
Pack
Volume (Metric):
3 mL
Quantity:
500/Pk., 3000/Cs.
Price (MYR)
640.00
Pack
Transfer liquids safely and accurately with Thermo Scientific™ Sterilin™ Plastic Transfer Pipettes. Ideal for general laboratory use, sample storage and transport.
Manufactured from nontoxic low density polyethylene
Integral filling bulb eliminates contamination associated with separate rubber bulbs
Excellent transparency and uniform thickness
Precise graduations on 1 and 3mL versions ensuring consistent results
Can be heat sealed for sample storage and transportation
Heat sealed bulbs suitable for storage in the gas phase of liquid nitrogen
3mL graduated: suitable for blood banking and general laboratory use
Narrow stem: suitable for use as a freeze vial, sedimentation and transport
1mL graduated: suitable for general laboratory use, storage and transportation