Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key
Actual product may vary
Fisherbrand
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key
For use when liquids have to be added to systems under vacuum.
Have Questions?
Catalog NumberCapacity (Metric)
FKS12905131250 mL
Catalog number FKS12905131
Price (MYR)
1,776.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (MYR)
1,776.00
EA
Add to cart
  • Complies with ISO 4800, Type 4
  • With interchangeable PTFE key for grease free performance
  • Double graduated scale showing volume of liquid contained/delivered
  • Stem inside cone for easy fitting to Quickfit™ assemblies
  • Graduations in white enamel
  • Supplied with hollow glass stopper
Specifications
Capacity (Metric)250 mL
MaterialBorosilicate Glass, Borosilicate Glass
GraduatedYes
Product TypePressure-equalizing Funnel
Unit SizeEA