Fisherbrand
Disposable Inoculation Loop, 10μL
Manufactured from polystyrene for strength and flexibility. Microspec™ Disposable Inoculation Loops are ideal for the cultivation of microbes on plates by transferring inoculum for streaking and to transfer microscopic organisms.
Have Questions?
Catalog NumberVolume
FKS1287015510 μL
Catalog number FKS12870155
Price (MYR)
503.00
Pack
Volume:
10 μL
Price (MYR)
503.00
Pack
  • High impact, polystyrene, disposable inoculating loops
  • 200mm long to reduce the risk of contamination
  • Aseptic Manufacture
Specifications
DisposableSingle-use
Length (Metric)200 mm
MaterialPlastic
SterilityAseptically Manufactured
Volume10 μL
Product TypeInoculating Loop
For Use With (Equipment)Laboratory and Research use
Quantity1000 Pk.
For Use With (Application)Laboratory and Research use
Unit SizePack