▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Brushes
▸
Beaker Brushes
Actual product may vary
Fisherbrand
Beaker Brushes
Constructed on galvanised steel wire handle. Pinnacle Brush™ Beaker Brushes have nylon bristles for gentle cleaning.
Have Questions?
Catalog Number
Length (Metric)
dimBristleMaterial
FKS12851934
36.5 cm
Catalog number
FKS12851934
Price (MYR)
297.00
Each
Add to cart
Length (Metric):
36.5 cm
Price (MYR)
297.00
Each
Add to cart
Galvanized steel handle
Nylon bristles
Specifications
Product Type
Bottle Brush
Handle Material
Metal
Diameter (Metric)
5.4 cm
Length (Metric)
36.5 cm
Length (Metric) Brush
12.5 cm
Length (Metric) Handle
24 cm
Quantity
10 Pk.
Size
Brush Circumference: 17 cm