PYREX™ Borosilicate Glass Graduated Cylinders
Actual product may vary
PYREX™ Borosilicate Glass Graduated Cylinders
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS12652042100 mL1 mL
Catalog number FKS12652042
Price (MYR)
1,277.00
Pack
Capacity (Metric):
100 mL
Graduations:
1 mL
Price (MYR)
1,277.00
Pack
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.
  • Hexagonal base delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
Specifications
Base TypeHexagonal
Scale ColorBlue
ShapeRound
TypeGraduated Cylinder
Tolerance±1 mL
MaterialBorosilicate Glass
Certifications/ComplianceBS 604, DIN 12680, ISO 4788
ClassB
Capacity (Metric)100 mL
Height (Metric)260 mm
ScaleBlue enamel
Graduations1 mL
Unit SizePack